Anti-Calcitonin Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-0124R-A750
BOSSBS-0124R-A750EA 874 EUR
BOSSBS-0124R-A750
Anti-Calcitonin Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
This is a receptor for calcitonin. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase. The calcitonin receptor is thought to couple to the heterotrimeric guanosine triphosphate-binding protein that is sensitive to cholera toxin.

Type: Primary
Antigen: Calcitonin receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-120/516
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR