Anti-Nogo Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-0129R-A750
BOSSBS-0129R-A750EA 890 EUR
BOSSBS-0129R-A750
Anti-Nogo Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Receptor for RTN4, OMG and MAG. Mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult central nervous system (By similarity). Acts in conjunction with RTN4 and LIGO1 in regulating neuronal precursor cell motility during cortical development.

Type: Primary
Antigen: Nogo Receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-350/473
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR