Anti-FGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-0229R-A680
BOSSBS-0229R-A680EA 890 EUR
BOSSBS-0229R-A680
Anti-FGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Plays an important role in the regulation of cell survival, cell division, angiogenesis, cell differentiation and cell migration. Functions as potent mitogen in vitro (By similarity).

Type: Primary
Antigen: FGF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 16-80/155
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR