Anti-EXPB2 Rabbit Polyclonal Antibody (Cy7®)

Proveedor: Bioss
BS-0243R-CY7
BOSSBS-0243R-CY7EA 900 EUR
BOSSBS-0243R-CY7
Anti-EXPB2 Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
beta-expansin (EXBP2) may cause loosening and extension of plant cell walls by disrupting noncovalent bonding between cellulose microfibrils and matrix glucans. No enzymatic activity has been found Subcellular Location at Cell-wall bound. Belongs to the expansin family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EXPB2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR