Anti-MAG Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-0257R-CY5
BOSSBS-0257R-CY5EA 909 EUR
BOSSBS-0257R-CY5
Anti-MAG Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Adhesion molecule in postnatal neural development that mediates sialic-acid dependent cell-cell interactions between neuronal and myelinating cells. Preferentially binds to alpha-2,3-linked sialic acid (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR