Anti-MAG Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-0258R-A750
BOSSBS-0258R-A750EA 890 EUR
BOSSBS-0258R-A750
Anti-MAG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Adhesion molecule in postnatal neural development that mediates sialic-acid dependent cell-cell interactions between neuronal and myelinating cells. Preferentially binds to alpha-2,3-linked sialic acid (By similarity).

Type: Primary
Antigen: MAG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-626/626
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR