Anti-PADI4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-0318R-A680
BOSSBS-0318R-A680EA 890 EUR
BOSSBS-0318R-A680
Anti-PADI4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
PADI4 (peptidyl arginine deiminase, type IV) catalyses the deimination of arginine residues of proteins. Down-regulates histone H3 and H4 arginine methylation, both by preventing arginine methylation by CARM1 and HRMT1L2/PRMT1 and by converting methylarginine to citrulline. subcellular location at cytoplasmic granules. Belongs to the protein arginine deiminase family. This gene may play a role in granulocyte and macrophage development leading to inflammation and immune response.

Type: Primary
Antigen: PADI4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR