Anti-HC Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Proveedor: Bioss
BS-0324R-A555
BOSSBS-0324R-A555EA 901 EUR
BOSSBS-0324R-A555
Anti-HC Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Activation of C5 by a C5 convertase initiates the spontaneous assembly of the late complement components, C5-C9, into the membrane attack complex. C5b has a transient binding site for C6. The C5b-C6 complex is the foundation upon which the lytic complex is assembled. Derived from proteolytic degradation of complement C5, C5 anaphylatoxin is a mediator of local inflammatory process. Binding to the receptor C5AR1 induces a variety of responses including intracellular calcium release, contraction of smooth muscle, increased vascular permeability, and histamine release from mast cells andMore Product Information