Anti-C5a anaphylatoxin Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-0324R-A680
BOSSBS-0324R-A680EA 899 EUR
BOSSBS-0324R-A680
Anti-C5a anaphylatoxin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Activation of C5 by a C5 convertase initiates the spontaneous assembly of the late complement components, C5-C9, into the membrane attack complex. C5b has a transient binding site for C6. The C5b-C6 complex is the foundation upon which the lytic complex is assembled. Derived from proteolytic degradation of complement C5, C5 anaphylatoxin is a mediator of local inflammatory process. Binding to the receptor C5AR1 induces a variety of responses including intracellular calcium release, contraction of smooth muscle, increased vascular permeability, and histamine release from mast cells andMore Product Information
Order Now

ESPECIFICACIONES

Nombre del antígeno C5a anaphylatoxin
Environmentally Preferable
Sinónimos de antígenos Hc|He|C5a|Hemolytic complement|Complement C5|C5
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 680
Reactividad Human,Mouse
Huésped Rabbit
ID del gen 15139
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Unmodified
Epítopo 701-755/1680
Inmunógeno KLH conjugated synthetic peptide derived from mouse C5a (725-770aa)
Forma Liquid
Molecular weight 188 kDa
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura de almacenamiento Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR