Anti-DLX4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-0352R-A488
BOSSBS-0352R-A488EA 893 EUR
BOSSBS-0352R-A488
Anti-DLX4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. Three transcript variants have been described for this gene, however, the full length nature of one variant hasMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno DLX4
Nombre del antígeno Distal-Less Homeobox 4
Environmentally Preferable
Sinónimos de antígenos DLX8|Beta protein 1 BP1|DLX7|Distal-Less Homeobox 7|DLX9|Distal-Less Homeobox 4 (isoform a)|BP1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 488
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 1748
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR