Anti-DLX4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: Bioss
BS-0352R-FITC
BOSSBS-0352R-FITCEA 900 EUR
BOSSBS-0352R-FITC
Anti-DLX4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Many vertebrate homeo box-containing genes have been identified on the basis of their sequence similarity with Drosophila developmental genes. Members of the Dlx gene family contain a homeobox that is related to that of Distal-less (Dll), a gene expressed in the head and limbs of the developing fruit fly. The Distal-less (Dlx) family of genes comprises at least 6 different members, DLX1-DLX6. The DLX proteins are postulated to play a role in forebrain and craniofacial development. Three transcript variants have been described for this gene, however, the full length nature of one variant hasMore Product Information