Anti-LDLR Rabbit Polyclonal Antibody (Cy7®)

Proveedor: Bioss
BS-0705R-CY7
BOSSBS-0705R-CY7EA 909 EUR
BOSSBS-0705R-CY7
Anti-LDLR Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
Binds LDL, the major cholesterol-carrying lipoprotein of plasma, and transports it into cells by endocytosis. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. In case of HIV-1 infection, functions as a receptor for extracellular Tat in neurons, mediating its internalization in uninfected cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LDLR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Other Species
Order Now

ESPECIFICACIONES

Simbolo antígeno LDLR
Nombre del antígeno Low-Density Lipoprotein Receptor
Environmentally Preferable
Sinónimos de antígenos FHC|FH|LDLCQ2|LDL receptor|LDL-R|Low-density lipoprotein receptor
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy7®
Reactividad Multiple,Human,Mouse
Huésped Rabbit
ID del gen 3949
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 811-850/860
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR