Anti-SPC Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-10067R-CY5
BOSSBS-10067R-CY5EA 909 EUR
BOSSBS-10067R-CY5
Anti-SPC Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Pulmonary surfactant associated proteins promote alveolar stability by lowering the surface tension at the air-liquid interface in the peripheral air spaces.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPC
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR