Anti-HINT1 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-10228R-CY3
BOSSBS-10228R-CY3EA 900 EUR
BOSSBS-10228R-CY3
Anti-HINT1 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
The protein encoded by this gene can hydrolyze substrates such as AMP-morpholidate, AMP-N-alanine methyl ester, AMP-alpha-acetyl lysine methyl ester, and AMP-NH2. The encoded protein interacts with these substrates via a histidine triad motif, which is part of the loop that binds to the substrate. This gene has been found to be a tumor suppressing gene. Several transcript variants, but only one of them protein-coding, have been found for this gene. [provided by RefSeq, Dec 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HINT1
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR