Anti-GLS Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-10341R-A555
BOSSBS-10341R-A555EA 893 EUR
BOSSBS-10341R-A555
Anti-GLS Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
This gene encodes the K-type mitochondrial glutaminase. The encoded protein is an phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. This protein is primarily expressed in the brain and kidney plays an essential role in generating energy for metabolism, synthesizing the brain neurotransmitter glutamate and maintaining acid-base balance in the kidney. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Jan 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLS1
Clonality: Polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR