Anti-F8 Rabbit Polyclonal Antibody
Proveedor: Bioss
BS-10397R
BOSSBS-10397REA 725 EUR
BOSSBS-10397R
Anti-F8 Rabbit Polyclonal Antibody
Anticuerpos
This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarilyMore Product Information