Anti-IFNLR1 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-11137R-CY3
BOSSBS-11137R-CY3EA 900 EUR
BOSSBS-11137R-CY3
Anti-IFNLR1 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
The IFNLR1/IL10RB dimer is a receptor for the cytokine ligands IFNL2 and IFNL3 and mediates their antiviral activity. The ligand/receptor complex stimulate the activation of the JAK/STAT signaling pathway leading to the expression of IFN-stimulated genes (ISG), which contribute to the antiviral state. Determines the cell type specificity of the lambda interferon action. Shows a more restricted pattern of expression in the epithelial tissues thereby limiting responses to lambda interferons primarily to epithelial cells of the respiratory, gastrointestinal, and reproductive tracts. Seems notMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno IFNLR1
Environmentally Preferable
Nombre del antígeno Interferon, Lambda Receptor 1
Sinónimos de antígenos CRF2/12|IL-28R1|LICR2|IL28RA|IFNLR
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy3®
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 163702
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 70-120/520
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR