Anti-RAB38 Rabbit Polyclonal Antibody
Proveedor: Bioss
BS-11244R
BOSSBS-11244REA 1090 EUR
BOSSBS-11244R
Anti-RAB38 Rabbit Polyclonal Antibody
Anticuerpos
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab superfamilies exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. The possiblity that Rab proteins might also directMore Product Information