Anti-PTPN5 Rabbit Polyclonal Antibody (Cy7®)
Proveedor: Bioss
BS-11328R-CY7
BOSSBS-11328R-CY7EA 900 EUR
BOSSBS-11328R-CY7
Anti-PTPN5 Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
The brain-specific STEP (striatal enriched phosphatase) family of protein tyrosine phosphatases (PTPs) comprises both transmembrane and cytosolic protein members which are the products of alternative splicing. STEP family members are expressed in the dopaminoceptive neurons of the CNS, with highest expression in the basal ganglia and related structures. The STEP protein regulates the N-methyl-d-aspartate receptor (NMDAR) complex; STEP depresses both NMDAR single-channel activity and synaptic currents. The membrane-associated STEP61 isoform localizes in the postsynaptic densities (PSDs) ofMore Product Information