Anti-GGACT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Bioss
BS-11381R-HRP
BOSSBS-11381R-HRPEA 900 EUR
BOSSBS-11381R-HRP
Anti-GGACT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Contributes to degradation of proteins cross-linked by transglutaminases. Degrades the cross-link between a lysine and a glutamic acid residue from two proteins that have been cross-linked by transglutaminases. Catalyzes the formation of 5-oxoproline from L-gamma-glutamyl-L-epsilon-lysine. Inactive with L-gamma-glutamyl-alpha-amino acid substrates such as L-gamma-glutamyl-L-alpha-cysteine and L-gamma-glutamyl-L-alpha-alanine.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: A2LD1/GGACT
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR