Anti-NPS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Bioss
BS-11429R-HRP
BOSSBS-11429R-HRPEA 1350 EUR
BOSSBS-11429R-HRP
Anti-NPS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Neuropeptides are regulators of synaptic transmission and their effects are mediated by G-protein coupled receptors. NPS (Neuropeptide S) is a 20 amino acid peptide cleaved from a larger precursor that contains a hydrophobic signal peptide and proteolytic cleavage processing sites. The N-terminal residue of NPS is always a serine regardless of the species. NPS is predominantly found in the central nervous system and plays an important role regulating sleep/wake functions, locomotion, arousal/anxiety responses and food intake. NPS functions by binding and activating its receptor, NPSR, andMore Product Information