Anti-SOX13 Rabbit Polyclonal Antibody (Cy5®)
Proveedor: Bioss
BS-11586R-CY5
BOSSBS-11586R-CY5EA 900 EUR
BOSSBS-11586R-CY5
Anti-SOX13 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of thisMore Product Information