Anti-PPP2R2B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-11749R-A488
BOSSBS-11749R-A488EA 893 EUR
BOSSBS-11749R-A488
Anti-PPP2R2B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) andMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno PPP2R2B
Environmentally Preferable
Nombre del antígeno Protein Phosphatase 2, Regulatory Subunit 2B
Sinónimos de antígenos Serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B beta isoform|PP2A subunit B isoform beta|PP2A PR55B|Protein phosphatase 2 regulatory subunit B|Serine/threonine protein phosphatase 2A neuronal isoform|PP2A subunit B isoform B55 beta|Protein phosphatase 2 regulatory subunit B beta isoform|PP2A subunit B PR55 beta isoform|PR2AB55 beta|PR2APR55 beta|SCA12|2ABB_HUMAN|Serine/Threonine Protein Phosphatase 2A 56kDa Regulatory Subunit Alpha Isoform|PP2A subunit B isoform B55-beta|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform.|Protein phosphatase 2 formerly 2A regulatory subunit B PR 52 beta isoform|PP2A B55beta|SCA 12|PP2A subunit B B beta isoform|Ppp2r2b|PP2A subunit B B55 beta isoform|Beta isoform of regulatory subunit B55 protein phosphatase 2|PP2A subunit B isoform R2 beta|PR52B|PP2A subunit B isoform PR55-beta|PP2A subunit B R2 beta isoform|PP2A subunit B isoform R2-beta|B55 beta|PR2AB beta|PP2A B Subunit B Alpha Isoform|PP2A subunit B isoform PR55 beta|MGC24888|PR55 beta|Protein phosphatase 2 formerly 2A regulatory subunit B beta isoform
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 488
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR