Anti-DNTNP/FAM53A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Proveedor: Bioss
BS-11839R-A750
BOSSBS-11839R-A750EA 890 EUR
BOSSBS-11839R-A750
Anti-DNTNP/FAM53A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Dorsal neural-tube nuclear protein is a 398 amino acid nuclear protein. DNTNP is thought to play an important role in neural development by specifying dorsal cell fates within the neural tube. Widely expressed in the dorsal neural tube, DNTNP is most highly expressed in the dorsal regions of the midbrain, the hindbrain, the diencephalon, and the spinal neural tube and is expressed at lower levels in the branchial arches, the telencephalon, the heart, and somites of developing embryos. DNTNP is encoded by a gene located on chromosome 4 which contains many genes including the Huntingtin gene,More Product Information