Anti-Radical Fringe Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Bioss
BS-11948R-HRP
BOSSBS-11948R-HRPEA 900 EUR
BOSSBS-11948R-HRP
Anti-Radical Fringe Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Three mammalian fringe family members, Manic, Radical and Lunatic Fringe, have been identified as proteins related to Drosophila Fringe, a protein involved in development. Fringe proteins act upstream of the Notch signaling pathway and are involved in boundary determination during segmentation. Each mammalian Fringe displays different patterns of expression, though all are expressed in the mouse embryo as well as in many adult tissues. Radical Fringe, also known as Beta-1,3-N-acetylglucosaminyltransferase Radical Fringe, is a 331 amino acid single-pass type II membrane protein that localizesMore Product Information