Anti-SLC25A12/ARALAR Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-11953R-A750
BOSSBS-11953R-A750EA 890 EUR
BOSSBS-11953R-A750
Anti-SLC25A12/ARALAR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Calcium Signalling in mitochondria is important in order for it to function in response to a variety of extracellular stimuli. Signalling begins with Ca²⁺ entry in mitochondria via the Ca²⁺ uniporter followed by Ca²⁺ activation of three dehydrogenases in the mitochondrial matrix. ARALAR, the neuronal Ca²⁺-binding mitochondrial aspartate-glutamate carrier, has Ca²⁺ binding domains facing the extramitochondrial space and functions in the malate-aspartate NADH shuttle (MAS). ARALAR is encoded by the SLC25A12 gene and is expressed in brain and skeletal muscle. ARALAR is required for theMore Product Information
Order Now

ESPECIFICACIONES

Nombre del antígeno SLC25A12/ARALAR
Environmentally Preferable
Sinónimos de antígenos ARALAR1|CMC1_HUMAN|solute carrier family 25, member 12|Araceli hiperlarga|Mitochondrial aspartate glutamate carrier 1|Calcium-binding mitochondrial carrier protein Aralar1|Solute carrier family 25 member 12|ARALAR|Calcium binding mitochondrial carrier superfamily member|AGC1|SLC25A12
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 750
Reactividad Mouse
Huésped Rabbit
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Unmodified
Epítopo 101-200/678
Inmunógeno KLH conjugated synthetic peptide derived from human SLC25A12/ARALAR
Forma Liquid
Molecular weight 75 kDa
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura de almacenamiento Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR