Anti-SNX7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Bioss
BS-12250R-HRP
BOSSBS-12250R-HRPEA 900 EUR
BOSSBS-12250R-HRP
Anti-SNX7 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomalMore Product Information