Anti-ZFP36L1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Proveedor: Bioss
BS-12254R-A488
BOSSBS-12254R-A488EA 893 EUR
BOSSBS-12254R-A488
Anti-ZFP36L1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
TIS11B is a member of the tristetraprolin family. Tristetraprolin (TTP), or TIS11, is a zinc-binding protein encoded by the immediate-early response gene, Zfp-36. TIS11B, a relative of TTP, localizes to the nucleus and may function as a transcription factor involved in regulating the growth factor response. It is an evolutionarily conserved protein containing two C3H1-type zinc fingers and a repeating cys-his motif. TIS11B is an mRNA binding protein and is known to interact with the 3’-untranslated region of VEGF mRNA, thereby decreasing its stability. This suggests that TIS11B is aMore Product Information