Anti-RBMY1F Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Bioss
BS-12290R-FITC
BOSSBS-12290R-FITCEA 900 EUR
BOSSBS-12290R-FITC
Anti-RBMY1F Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
The RBM (RNA-binding motif) gene family encodes proteins with an RNA binding motif. RBMY (RBM, Y chromosome) encodes a germ-cell specific nuclear protein involved in spermatogenesis. The RBM gene family, including RBMY1A, RBMY1B, RBMY1D, RBMY1E, RBMY1F, RBMY1H and RBMY1J, is comprised of at least 30 genes and pseudogenes, found on both arms of the Y chromosome. RBM X, an ancestral X chromosome homolog of the RBMY gene, encodes hnRNP G, which is widely expressed, whereas the RBMY gene evolved a male-specific function in spermatogenesis. Micro-deletions of the AZFb region of the Y chromosome,More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno RBMY1F
Nombre del antígeno RNA Binding Motif Protein Y Linked, Family 1, Member F
Environmentally Preferable
Sinónimos de antígenos YRRM2|RBY1F_HUMAN.|member F|family 1|Y chromosome|Y chromosome RNA recognition mot 2|family 1 member F/J|RNA binding mot protein|Y linked
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human
Huésped Rabbit
ID del gen 159163
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR