Anti-VPREB1 Rabbit Polyclonal Antibody
Proveedor: Bioss
BS-12326R
BOSSBS-12326REA 725 EUR
BOSSBS-12326R
Anti-VPREB1 Rabbit Polyclonal Antibody
Anticuerpos
VpreB is expressed in pre-B lymphocytes, but not in mature B cells or in other blood cell lineages. The gene which encodes VpreB maps to human chromosome 22q11.2. The VpreB and Lambda 5 genes encode the Iota and Omega polypeptide chains, respectively, which associate with the Ig-Mu chain to form a molecular complex that is expressed on the surface of pre-B cells. This complex presumably regulates Ig gene rearrangements in the early steps of B cell differentiation. In the mouse the two genes are simultaneously expressed in pre-B cells and belong to the same transcription unit. A primaryMore Product Information