Anti-MIXL1 Rabbit Polyclonal Antibody (Cy3®)
Proveedor: Bioss
BS-12350R-CY3
BOSSBS-12350R-CY3EA 900 EUR
BOSSBS-12350R-CY3
Anti-MIXL1 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
The homeobox DNA-binding domain is a 60 amino acid motif that is conserved among many species and functions to bind DNA via a helix-turn-helix structure, thereby playing a role in transcriptional regulation and in the control of gene expression. MIXL1 (Mix1 homeobox-like 1), also known as MIXL, is a 232 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. Expressed in lymph tissues, MIXL1 functions as a transcription factor that plays an essential role in axial mesendoderm morphogenesis and endoderm formation and is also required for cellularMore Product Information