Anti-HYAL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-1235R-A488
BOSSBS-1235R-A488EA 893 EUR
BOSSBS-1235R-A488
Anti-HYAL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
May have a role in promoting tumor progression. May block the TGFB1-enhanced cell growth (By similarity). Overexpression of HYAL1 suppressed the growth rate of colon carcinoma cell tumors in an experimental model.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HYAL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno HYAL1
Environmentally Preferable
Nombre del antígeno Hyaluronidase 1
Sinónimos de antígenos HYAL-1|NAT6|MPS9|LUCA1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 488
Reactividad Rat,Mouse
Huésped Rabbit
ID del gen 367166
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 270-320/449
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR