Anti-ARL4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Proveedor: Bioss
BS-12523R-A350
BOSSBS-12523R-A350EA 893 EUR
BOSSBS-12523R-A350
Anti-ARL4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
ADP-ribosylation factors (ARFs) are highly conserved guanine nucleotide-binding proteins that enhance the ADP-ribosyltransferase activity of cholera toxin. ARF抯 are important in eukaryotic vesicular trafficking pathways and activating phospholipase D. ARL4 (ADP-ribosylation factor-like protein 4A) is a member of the ARF-like protein (ARL) subfamily of small GTPases. It contains a C terminal nuclear localization signal (NLS) region that interacts with Importin-? ARL4 localizes to the nucleus and is found in a variety of tissues, but is predominantly expressed in spermatogonia and SertoliMore Product Information