Anti-BOLL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: Bioss
BS-12880R-FITC
BOSSBS-12880R-FITCEA 900 EUR
BOSSBS-12880R-FITC
Anti-BOLL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Spermatogenesis represents the intricate developmental process of mitotic and meiotic cell divisions that ultimately leads to the production of haploid spermatozoa. BOULE, a member of the human deleted in azoospermia (DAZ) family, functions as a key conserved switch that regulates the progression of germ cells through meiosis in man. BOULE is an RNA-binding protein that regulates the expression of twine, a Cdc25 phosphatase, which promotes progression through meiosis. BOULE is expressed not only in the testis, but also in the nervous system, where it may play a role in neural communication.More Product Information