Anti-REL Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Proveedor: Bioss
BS-12918R-A647
BOSSBS-12918R-A647EA 898 EUR
BOSSBS-12918R-A647
Anti-REL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticuerpos
The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA (MIM 164014), RELB (604758), NFKB1 (MIM 164011), and NFKB2 (MIM 164012). These proteins are related through a highly conserved N-terminal region termed the 'Rel domain,' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (MIM 164008) (Belguise and Sonenshein, 2007 [PubMed 18037997]).[supplied by OMIM, May 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: REL
Clonality: Polyclonal
Clone:
More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno REL
Nombre del antígeno V-rel Avian Reticuloendotheliosis Viral Oncogene homolog
Environmentally Preferable
Sinónimos de antígenos 4.1-G|c-Rel|4.1G
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 647
Reactividad Human,Mouse
Huésped Rabbit
ID del gen 5966
Isotipo IgG
InmunoQuímica Yes
Modificación phospho-Ser503
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR