Anti-POLK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: Bioss
BS-13019R-FITC
BOSSBS-13019R-FITCEA 900 EUR
BOSSBS-13019R-FITC
Anti-POLK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
DNA polymerase lambda (pol Kappa), also designated DNA polymerase Kappa or Pol Beta2, is a low-fidelity polymerase which plays a role in both spontaneous and DNA damage-induced mutagenesis. Encoded by the POLL gene, pol Kappa is a member of the DNA polymerase type-X family. Pol Kappa extends primer-terminal mispairs opposite nondamaged DNA templates, suggesting that it may assist in extending mismatched base pairs during normal DNA replication. In addition, pol Kappa may play a role in the mutagenic bypass of T-T dimers. Proliferating cell nuclear antigen (PCNA), a protein essential to DNAMore Product Information