Anti-FAM134B Rabbit Polyclonal Antibody (Cy7®)

Proveedor: Bioss
BS-13136R-CY7
BOSSBS-13136R-CY7EA 900 EUR
BOSSBS-13136R-CY7
Anti-FAM134B Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
Required for long-term survival of nociceptive and autonomic ganglion neurons.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM134B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR