Anti-FUOM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Bioss
BS-13226R-HRP
BOSSBS-13226R-HRPEA 1350 EUR
BOSSBS-13226R-HRP
Anti-FUOM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Fukutin, a secreted protein, is expressed in various tissues in normal individuals. Fukutin colocalizes with a Golgi marker and a granular cytoplasmic distribution, suggesting that fukutin passes through the Golgi before being packaged into secretory vesicles. Fukutin may be located in the extracellular matrix, where it interacts with and reinforces a large complex encompassing the outside and inside of muscle membranes; alternatively, as a secreted protein, fukutin may cause muscular dystrophy by an unknown mechanism. The fukutin gene is expressed at similar levels in control fetal andMore Product Information