Anti-GBA3/CBG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Proveedor: Bioss
BS-13298R-A750
BOSSBS-13298R-A750EA 890 EUR
BOSSBS-13298R-A750
Anti-GBA3/CBG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
CBG is a monomeric enzyme involved in the absorption and metabolism of flavonoid glucosides. CBG is found predominately in the liver, but is also located in tissues such as spleen, small intestine and kidney. Through its catalytic activity, CBG is able to hydrolyze a variety of glycosides including phytoestrogens, cyanogens, and flavonols. Although its catalytic activity extends to many dietary flavonoids, CBG has increased specificity for hydrophobic aglycones such as beta-D-glucoside and beta-D-galactoside. Hydrolysis is inhibited by sodium taurocholate and glucosyl-sphingosine, both ofMore Product Information