Anti-GLIPR1L1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: Bioss
BS-13375R-FITC
BOSSBS-13375R-FITCEA 900 EUR
BOSSBS-13375R-FITC
Anti-GLIPR1L1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
The GLIPR1 family consists of three core members, designated GLIPR1, GLIPR1L1 (GLIPR1-like protein 1) and GLIPR1L2, which form a distinct subgroup within the cysteine-rich secretory protein (CRISP), antigen 5 and pathogenesis-related 1 (CAP) superfamily. Each member of the CAP superfamily has a conserved N-terminal CAP domain and a distinct C-terminal extension. CAP superfamily proteins are hypothesized to have roles in immunity, cell adhesion, carcinogenesis and male fertility. GLIPR1L1 is a 242 amino acid secreted protein. Highly expressed in testis, GLIPR1L1 exists as two isoformsMore Product Information