Anti-NRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-1342R-A488
BOSSBS-1342R-A488EA 901 EUR
BOSSBS-1342R-A488
Anti-NRF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Transcription factor that activates the expression of the EIF2S1 (EIF2-alpha) gene. Links the transcriptional modulation of key metabolic genes to cellular growth and development. Implicated in the control of nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NRF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR