Anti-CLCNKB Rabbit Polyclonal Antibody
Proveedor: Bioss
BS-13627R
BOSSBS-13627REA 725 EUR
BOSSBS-13627R
Anti-CLCNKB Rabbit Polyclonal Antibody
Anticuerpos
The family of voltage-dependent chloride channels (CLCs) regulate cellular trafficking of chloride ions, a critical component of all living cells. CLCs regulate excitability in muscle and nerve cells, aid in organic solute transport, and maintain cellular volume. CLC-KA is a kidney-specific chloride channel that mediates transepithelial chloride transport in the thin ascending limb of the Henle loop in the inner medulla. CLC-KA plays a crucial role in urine concentration. The gene encoding human CLC-KA maps to chromosome 1p36. Mutations in this gene may be associated with nephrogenicMore Product Information