Anti-AIF1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Bioss
BS-1363R-HRP
BOSSBS-1363R-HRPEA 909 EUR
BOSSBS-1363R-HRP
Anti-AIF1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Actin-binding protein that enhances membrane ruffling and RAC activation. Enhances the actin-bundling activity of LCP1. Binds calcium. Plays a role in RAC signaling and in phagocytosis. May play a role in macrophage activation and function. Promotes the proliferation of vascular smooth muscle cells and of T-lymphocytes. Enhances lymphocyte migration. Plays a role in vascular inflammation.

Recommended Dilutions: IHC-P: 1:100-500; IHC-F(1:100-500

Type: Primary
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR