Anti-Claudin 16 Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-13752R-CY5
BOSSBS-13752R-CY5EA 900 EUR
BOSSBS-13752R-CY5
Anti-Claudin 16 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity. Involved in paracellular magnesium reabsorption. Required for a selective paracellular conductance. May form, alone or in partnership with other constituents, an intercellular pore permitting paracellular passage of magnesium and calcium ions down their electrochemical gradients. Alternatively, it could be a sensor of magnesium concentration that could alter paracellular permeability mediated by other factors.

Type: Primary
Antigen: Claudin 16
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR