Anti-Arfaptin 2 Rabbit Polyclonal Antibody (Cy5.5®)

Proveedor: Bioss
BS-1442R-CY5.5
BOSSBS-1442R-CY5.5EA 900 EUR
BOSSBS-1442R-CY5.5
Anti-Arfaptin 2 Rabbit Polyclonal Antibody (Cy5.5®)
Anticuerpos
ARFIP2 is a ubiquitously expressed protein implicated in mediating cross talk between RAC and ARF small GTPases. It has been shown that ARFIP2 binds specifically to GTP-bound ARF1 and ARF6, but binds to Rac-GTP and Rac-GDP with similar affinities. The X-ray structure of arfaptin reveals an elongated, crescent-shaped dimer of 3-helix coiled-coils. Structures of arfaptin with Rac bound to either GDP or the slowly hydrolysable analog GMPPNP show that the switch regions adopt similar conformations in both complexes.

Type: Primary
Antigen: Arfaptin 2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR