Anti-Promotilin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Bioss
BS-1445R-FITC
BOSSBS-1445R-FITCEA 900 EUR
BOSSBS-1445R-FITC
Anti-Promotilin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Plays an important role in the regulation of interdigestive gastrointestinal motility and indirectly causes rhythmic contraction of duodenal and colonic smooth muscle.

Type: Primary
Antigen: Promotilin
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 51-115/115
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR