Anti-C20orf117 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-15091R-A555
BOSSBS-15091R-A555EA 893 EUR
BOSSBS-15091R-A555
Anti-C20orf117 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 2More Product Information
Order Now

ESPECIFICACIONES

Environmentally Preferable
Nombre del antígeno C20orf117
Sinónimos de antígenos CT117_HUMAN|C20orf117|hypothetical protein LOC140710|dJ132F21.1|KIAA0889|Uncharacterized protein C20orf117|FLJ44670
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 555
Reactividad Mouse
Huésped Rabbit
ID del gen 140710
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Unmodified
Epítopo 21-120/1423
Inmunógeno KLH conjugated synthetic peptide derived from human C20orf117
Forma Liquid
Molecular weight 160 kDa
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Temperatura de almacenamiento Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR