Anti-GPR158 Rabbit Polyclonal Antibody (HRP)

Proveedor: Bioss
BS-15384R-HRP
BOSSBS-15384R-HRPEA 900 EUR
BOSSBS-15384R-HRP
Anti-GPR158 Rabbit Polyclonal Antibody (HRP)
Anticuerpos
GPR158.

Type: Primary
Antigen: GPR158
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 201-300/1215
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR