Anti-CYP51A1 Rabbit Polyclonal Antibody (Cy3)

Proveedor: Bioss
BS-15409R-CY3
BOSSBS-15409R-CY3EA 900 EUR
BOSSBS-15409R-CY3
Anti-CYP51A1 Rabbit Polyclonal Antibody (Cy3)
Anticuerpos
CYP51A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyse many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein participates in the synthesis of cholesterol by catalysing the removal of the 14alpha-methyl group from lanosterol. Homologous genes are found in all three eukaryotic phyla, fungi, plants, and animals, suggesting that this is one of the oldest cytochrome P450 genes. Two transcript variants encoding different isoforms have beenMore Product Information
Order Now

ESPECIFICACIONES

Environmentally Preferable
Nombre del antígeno CYP51A1
Sinónimos de antígenos Cytochrome P450-14DM|Cytochrome P450LI|CP51A_HUMAN|Cytochrome P45014DM|Sterol 14-alpha demethylase|LDM|Lanosterol 14-alpha demethylase|Cytochrome P450 51A1|CYPLI
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy3®
Reactividad
Huésped Rabbit
ID del gen 1595
Isotipo IgG
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Unmodified
Epítopo 411-503/503
Inmunógeno KLH conjugated synthetic peptide derived from human CYP51A1
Forma Liquid
Molecular weight 57 kDa
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura de almacenamiento Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR