Anti-IFIT1B Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)

Proveedor: Bioss
BS-15554R-A350
BOSSBS-15554R-A350EA 893 EUR
BOSSBS-15554R-A350
Anti-IFIT1B Rabbit Polyclonal Antibody (ALEXA FLUOR® 350)
Anticuerpos
IFIT1B.

Type: Primary
Antigen: IFIT1B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 350
Public Immunogen Range: 351-474/474
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR